2-Amino-4-(trifluormethyl)benzenthiol hydrochlorid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Amino-4-(trifluoromethyl)benzenethiol hydrochloride
M-Clarity™ quality level = MQ100
365734-25G
365734-5G
SIAL365734-25GEA
12140
CZK
SIAL365734-25G
SIAL365734-5G
2-Amino-4-(trifluormethyl)benzenthiol hydrochlorid, Sigma-Aldrich®
2-Amino-4-(trifluormethyl)benzenthiol hydrochlorid
Vzorec:
C₇H₇ClF₃NS |
MDL Number:
MFCD00042600 CAS číslo: 4274-38-8 EINECS: 429-560-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...