Benzofenon-3,4,3´,4´-tetrakarboxdianhydrid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4,4'-Karbonylbis(ftalanhydrid)
M-Clarity™ quality level = MQ200
B9750-100G
B9750-500G
SIALB9750-100GEA
1560
CZK
SIALB9750-100G
SIALB9750-500G
Benzofenon-3,4,3´,4´-tetrakarboxdianhydrid, Sigma-Aldrich®
Benzofenon-3,4,3´,4´-tetrakarboxdianhydrid
Vzorec:
C₁₇H₆O₇ |
MDL Number:
MFCD00005923 CAS číslo: 2421-28-5 EINECS: 219-348-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...