2-Chloro-6-fluorobenzaldehyde oxime ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Chloro-6-fluorobenzaldoxime
L14719.03
L14719.06
L14719.03EA
570
CZK
L14719.03
L14719.06
2-Chloro-6-fluorobenzaldehyde oxime ≥97%
2-Chloro-6-fluorobenzaldehyde oxime
Purity: 97%
CAS No.: 443-33-4
Molecular Formula: C7H5ClFNO
Formula Weight: 173.58
Melting Point: 125-127°
Beilstein Reference: 3247886
Vzorec:
C₇H₅ClFNO MW: 173,57 g/mol Bod tání: 125…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00837704 CAS číslo: 443-33-4 EINECS: 207-135-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...