Tisk…

4-Bromo-3-nitrobenzaldehyde, Sigma-Aldrich®

Dodavatel: Merck

523151-1G 523151-5G
SIAL523151-1GEA 848 CZK
SIAL523151-1G SIAL523151-5G
4-Bromo-3-nitrobenzaldehyde, Sigma-Aldrich®
4-Bromo-3-nitrobenzaldehyde
Vzorec: C₇H₄BrNO₃
MW: 230,02 g/mol
MDL Number: MFCD00204031
CAS číslo: 163596-75-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR