Tisk…

Erythrosin B disodium salt

Dodavatel: Thermo Fisher Scientific
Varování

A14180.14 A14180.22 A14180.36
A14180.14EA 1490 CZK
A14180.14 A14180.22 A14180.36
Erythrosin B disodium salt
Erythrosin B disodium salt

CAS No.: 568-63-8 Molecular Formula: C20H6I4Na2O5 Formula Weight: 879.87 MDL No.: MFCD00144257


Acid Red 51

Vzorec: C₂₀H₆I₄Na₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00144257
CAS číslo: 16423-68-0
EINECS: 240-046-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR