Tisk…

1-Ethyl-3-methylimidazolium diethylphosphate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-ethyl-3-methylimidazolium diethylphosphate

713392-100G 713392-1KG
SIAL713392-100GEA 8880 CZK
SIAL713392-100G SIAL713392-1KG
1-Ethyl-3-methylimidazolium diethylphosphate, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium diethylphosphate
Vzorec: C₁₀H₂₁N₂O₄P
MW: 264,26 g/mol
MDL Number: MFCD09953486
CAS číslo: 848641-69-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR