1-Butyl-3-methylimidazolium acetate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
30367-50G
30367-5G
SIAL30367-50GEA
25940
CZK
SIAL30367-50G
SIAL30367-5G
1-Butyl-3-methylimidazolium acetate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium acetate
Vzorec:
C₁₀H₁₈N₂O₂ MW: 198,27 g/mol |
MDL Number:
MFCD06798175 CAS číslo: 284049-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...