1-Boc-indole-2-boronic acid 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acid, 1H-Indole-2-boronic acid, N-BOC protected, N-Boc-indole-2-boronic Acid, 1-(tert-Butoxycarbonyl)indole-2-boronic acid, N-Boc-indole-2-boronic acid
L18009.03
L18009.06
L18009.MD
L18009.03EA
3710
CZK
L18009.03
L18009.06
L18009.MD
1-Boc-indole-2-boronic acid 95%
1-Boc-indole-2-boronic acid
Purity: 95%
CAS No.: 213318-44-6
Molecular Formula: C13H16BNO4
Formula Weight: 261.08
Storage Temperature: 2°C to 8°C
Melting Point: ca 85° dec.
Beilstein Reference: 8148683
MDL No.: MFCD02093045
Vzorec:
C₁₃H₁₆BNO₄ MW: 261,08 g/mol Bod tání: ∼ 85 °C (rozkl.) |
MDL Number:
MFCD02093045 CAS číslo: 213318-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...