Tisk…

3-Amino-4-bromobenzoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Amino-4-bromobenzoic Acid

M-Clarity™ quality level = MQ100

686433-25G 686433-5G
SIAL686433-25GEA 12660 CZK
SIAL686433-25G SIAL686433-5G
3-Amino-4-bromobenzoic acid, Sigma-Aldrich®
3-Amino-4-bromobenzoic acid
Vzorec: C₇H₆BrNO₂
MW: 216,03 g/mol
MDL Number: MFCD00579106
CAS číslo: 2840-29-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR