Tisk…

4-Bromo-o-anisaldehyde, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Bromo-2-methoxybenzaldehyde

M-Clarity™ quality level = MQ100

661880-5G 661880-1G
SIAL661880-5GEA 0 CZK
SIAL661880-5G SIAL661880-1G
4-Bromo-o-anisaldehyde, Sigma-Aldrich®
4-Bromo-o-anisaldehyde
Vzorec: C₈H₇BrO₂
MW: 215,05 g/mol
MDL Number: MFCD08276823
CAS číslo: 43192-33-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR