6-Bromo-m-anisaldehyde, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Bromo-5-methoxybenzaldehyde
702390-1G
702390-5G
SIAL702390-1GEA
1180
CZK
SIAL702390-1G
SIAL702390-5G
6-Bromo-m-anisaldehyde, Sigma-Aldrich®
6-Bromo-m-anisaldehyde
Vzorec:
C₈H₇BrO₂ MW: 215,05 g/mol |
MDL Number:
MFCD00463817 CAS číslo: 7507-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...