Tisk…

Isoamyl butyrate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Isopentyl butyrate, Isoamyl Butyrate

B21103.22 B21103.36
B21103.22EA 1020 CZK
B21103.22 B21103.36
Isoamyl butyrate ≥98%
Isoamyl butyrate

Purity: 98% CAS No.: 106-27-4 Molecular Formula: CH3CH2CH2CO2CH2CH2CH(CH3)2 Formula Weight: 158.24 Flash Point: 59°(138°F) Refractive Index: 1.411 Odour: Fruit-like Melting Point: -73° Boiling Point: 178° Merck Reference: 14,5115 UN No.: 2620 MDL No.: MFCD00044888

Vzorec: C₉H₁₈O₂
Bod varu: 178 °C
Bod tání: –73 °C
Hustota: 0,88
Bod vzplanutí: 59 °C (138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044888
CAS číslo: 106-27-4
EINECS: 203-380-8
UN: 2620
ADR: 3,III
Merck Index: 14,5115

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR