Tisk…

Diglycolic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

143073-100G 143073-1KG 143073-5G
SIAL143073-100GEA 2550 CZK
SIAL143073-100G SIAL143073-1KG SIAL143073-5G
Diglycolic acid, Sigma-Aldrich®
Diglycolic acid
Vzorec: C₄H₆O₅
MDL Number: MFCD00004309
CAS číslo: 110-99-6
EINECS: 203-823-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR