Tisk…

1-Methyl-3-octylimidazolium hexafluorophosphate, Sigma-Aldrich®

Dodavatel: Merck

69230-25G-F 69230-5G-F
SIAL69230-25G-FEA 9810 CZK
SIAL69230-25G-F SIAL69230-5G-F
1-Methyl-3-octylimidazolium hexafluorophosphate, Sigma-Aldrich®
1-Methyl-3-octylimidazolium hexafluorophosphate
Vzorec: C₁₂H₂₃F₆N₂P
MW: 340,29 g/mol
MDL Number: MFCD06798203
CAS číslo: 304680-36-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR