Tisk…

6-Methoxytryptamine 99%

Dodavatel: Thermo Scientific
Nebezpečí

187452500 187450010
ACRO187452500EA 7140 CZK
ACRO187452500 ACRO187450010
6-Methoxytryptamine 99%
6-Methoxytryptamine
Vzorec: C₁₁H₁₄N₂O
Bod tání: 143…146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005663
CAS číslo: 3610-36-4
EINECS: 222-778-2
UN: 3259
ADR: 8,III

Objednat

Výsledky specifikačního testu

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 146 °C
Water ≤1 % (K.F.)
Appearance of solution (2% soln. in ethanol) practically clear, almost colorless to yellowish
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR