Tisk…

5-Methoxytryptamine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

286583-100MG 286583-1G
SIAL286583-100MGEA 1270 CZK
SIAL286583-100MG SIAL286583-1G
5-Methoxytryptamine, Sigma-Aldrich®
5-Methoxytryptamine
Vzorec: C₁₁H₁₄N₂O
MDL Number: MFCD00005662
CAS číslo: 608-07-1
EINECS: 210-153-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR