Tisk…

1-Ethyl-3-methylimidazolium iodide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-ethyl-3-methylimidazolium iodide

713031-50G 713031-5G
SIAL713031-50GEA 9430 CZK
SIAL713031-50G SIAL713031-5G
1-Ethyl-3-methylimidazolium iodide, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium iodide
Vzorec: C₆H₁₁IN₂
MW: 238,07 g/mol
MDL Number: MFCD03701101
CAS číslo: 35935-34-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR