Tisk…

Disodium malonate monohydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium malonate dibasic monohydrate, Malonic acid disodium salt hydrate

M-Clarity™ quality level = MQ100

M4795-10G M4795-25G
SIALM4795-10GEA 2530 CZK
SIALM4795-10G SIALM4795-25G
Disodium malonate monohydrate, Sigma-Aldrich®
Disodium malonate monohydrate
Vzorec: C₃H₂Na₂O₄·1H₂O
MW: 166,04 g/mol
MDL Number: MFCD00149114
CAS číslo: 26522-85-0
EINECS: 205-514-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR