Tisk…

Disodium malonate monohydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium malonate dibasic monohydrate, Malonic acid disodium salt hydrate

M1875-100G M1875-25G M1875-500G
SIALM1875-100GEA 3590 CZK
SIALM1875-100G SIALM1875-25G SIALM1875-500G
Disodium malonate monohydrate, Sigma-Aldrich®
Disodium malonate monohydrate
Vzorec: C₃H₂Na₂O₄·1H₂O
MW: 166,04 g/mol
MDL Number: MFCD00149114
CAS číslo: 26522-85-0
EINECS: 205-514-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR