Tisk…

1-Methyl-3-octylimidazolium tetrafluoroborate ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: 1-Methyl-3-n-octylimidazolium Tetrafluoroborate, 1-octyl-3-methylimidazolium tetrafluoroborate, [OMIM] [BF4]

H27161.06 H27161.18
H27161.06EA 2950 CZK
H27161.06 H27161.18
1-Methyl-3-octylimidazolium tetrafluoroborate ≥99%
1-Methyl-3-octylimidazolium tetrafluoroborate

Purity: 99% CAS No.: 244193-52-0 Molecular Formula: C12H23BF4N2 Formula Weight: 282.13 MDL No.: MFCD03095434

Vzorec: C₁₂H₂₃BF₄N₂
Hustota: 1,120
Storage Temperature: Ambient
MDL Number: MFCD03095434
CAS číslo: 244193-52-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR