Tisk…

(±)-Methyl-2-methylbutyrate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Methyl2-methylbutyrate

L08103.14 L08103.22
L08103.14EA 982 CZK
L08103.14 L08103.22
(±)-Methyl-2-methylbutyrate ≥98%
(±)-Methyl-2-methylbutyrate

Purity: 98% CAS No.: 868-57-5 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 16°(61°F) Refractive Index: 1.3940 Odour: Fruit-like Boiling Point: 114-116° Beilstein Reference: 1720409 UN No.: 3272 MDL No.: MFCD00009335

Vzorec: C₆H₁₂O₂
MW: 116.16
Bod varu: 114…116 °C
Hustota: 0.885
Bod vzplanutí: 16 °C (61 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009335
CAS číslo: 868-57-5
EINECS: 212-778-0
UN: 3272
ADR: 3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR