Tisk…

Terbium triacetate hydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Terbium(III) acetate hydrate

M-Clarity™ quality level = MQ100

325929-25G 325929-5G
SIAL325929-25GEA 19640 CZK
SIAL325929-25G SIAL325929-5G
Terbium triacetate hydrate, Sigma-Aldrich®
Terbium triacetate hydrate
Vzorec: C₆H₉O₆Tb·H₂O
MW: 354.07
MDL Number: MFCD00149233
CAS číslo: 100587-92-6
EINECS: 247-065-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR