Tisk…

Erbium triacetate tetrahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Fisher Scientific

Synonyms: Erbium(III) acetate tetrahydrate

40481.14 40481.06 40481.22
40481.14EA 6490 CZK
40481.14 40481.06 40481.22
Erbium triacetate tetrahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium triacetate tetrahydrate

Purity: 99.9% CAS No.: 15280-57-6 Molecular Formula: C6H9ErO6.4H2O Formula Weight: 416.45 (344.39anhy) Physical Form: Crystalline Aggregates Density: 2.114 MDL No.: MFCD00013034

Vzorec: C₆H₉ErO₆·4H₂O
MW: 416.45
Hustota: 2.114
Storage Temperature: Ambient
MDL Number: MFCD00013034
CAS číslo: 15280-57-6
EINECS: 247-067-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR