Methyl isobutyrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
148008-100ML
148008-500ML
SIAL148008-100MLEA
1820
CZK
SIAL148008-100ML
SIAL148008-500ML
Methyl isobutyrate, Sigma-Aldrich®
Methyl isobutyrate
Vzorec:
C₅H₁₀O₂ MW: 102,13 g/mol |
MDL Number:
MFCD00008914 CAS číslo: 547-63-7 EINECS: 208-929-5 UN: 1237 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...