Tisk…

Isobutyl isobutyrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W218901-1KG-K W218901-4KG-K
SIALW218901-1KG-KEA 2630 CZK
SIALW218901-1KG-K SIALW218901-4KG-K
Isobutyl isobutyrate, Sigma-Aldrich®
Isobutyl isobutyrate
Vzorec: C₈H₁₆O₂
MW: 144,21 g/mol
MDL Number: MFCD00008916
CAS číslo: 97-85-8
EINECS: 202-612-5
UN: 2528
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR