L-allo-Threonine 99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
H-allo-Thr-OH, L(+)-allo-Threonine, Allo-Thr-OH
198541000
198542500
ACRO198541000EA
1800
CZK
ACRO198541000
ACRO198542500
L-allo-Threonine 99%
L-allo-Threonine
Vzorec:
C₄H₉NO₃ MW: 119.12 Bod tání: 256 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00064268 CAS číslo: 28954-12-3 EINECS: 249-327-2 |
Výsledky specifikačního testu
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Specific optical rotation | +8° to +10° (20°C, 589 nm) (c=2, water) |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...