L-Threonine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
M-Clarity™ quality level = MQ200
8.16012.0025
8.16012.0100
8.16012.0025EA
1950
CZK
8.16012.0025
8.16012.0100
L-Threonine, Sigma-Aldrich®
L-Threonine
Vzorec:
C₄H₉NO₃ MW: 119,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064270 CAS číslo: 72-19-5 EINECS: 200-774-1 |
Výsledky specifikačního testu
Solubility | 90 g/l (20 °C) |
Melting Point | 265 - 270 °C |
Molar Mass | 119.12 g/mol |
Bulk Density | 370 kg/m3 |
pH | 5 - 6 (100 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...