Tisk…

(S)-(+)-2-Aminobutyric acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: H-Abu-OH, L-alpha-Amino-N-Butyric acid, L-2-Aminobutyric acid, (S)-2-Aminobutanoic acid

L13984.03 L13984.06 L13984.14
L13984.03EA 1740 CZK
L13984.03 L13984.06 L13984.14
(S)-(+)-2-Aminobutyric acid ≥98%
(S)-(+)-2-Aminobutyric acid

Purity: 98+% CAS No.: 1492-24-6 Molecular Formula: C4H9NO2 Formula Weight: 103.12 Melting Point: 300° Beilstein Reference: 1720935 Merck Reference: 14,428 Optical Rotation: +21° (c=4.8 in 6N HCl) MDL No.: MFCD00064415

Vzorec: C₄H₉NO₂
MW: 103,12 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064415
CAS číslo: 1492-24-6
EINECS: 216-083-3
Merck Index: 14,428

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR