Tisk…

(S)-(+)-2-Aminobutyric acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ400

A1879-1G A1879-5G
SIALA1879-1GEA 5060 CZK
SIALA1879-1G SIALA1879-5G
(S)-(+)-2-Aminobutyric acid, Sigma-Aldrich®
(S)-(+)-2-Aminobutyric acid
Vzorec: C₄H₉NO₂
MDL Number: MFCD00064415
CAS číslo: 1492-24-6
EINECS: 216-083-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR