Tisk…

(R)-(-)-2-Aminobutyric acid ≥98%

Dodavatel: Thermo Scientific

Synonyms: (R)-()-2-Aminobutyric acid, D-2-Aminobutyric acid, D(-)-2-Aminobutyric acid, H-D-Abu-OH

L14096.03 L14096.06
L14096.03EA 1380 CZK
L14096.03 L14096.06
(R)-(-)-2-Aminobutyric acid ≥98%
(R)-(-)-2-Aminobutyric acid

Purity: 98+% CAS No.: 2623-91-8 Molecular Formula: C4H9NO2 Formula Weight: 103.12 Melting Point: 300° Beilstein Reference: 1720934

Vzorec: C₄H₉NO₂
MW: 103,12 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064414
CAS číslo: 2623-91-8
EINECS: 220-084-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR