Tisk…

(±)-2-Aminobutyric acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: H-DL-Abu-OH, DL-2-Aminobutyric acid, DL-α-Amino-N-Butyric Acid

M-Clarity™ quality level = MQ200

162663-100G 162663-25G
SIAL162663-100GEA 3640 CZK
SIAL162663-100G SIAL162663-25G
(±)-2-Aminobutyric acid, Sigma-Aldrich®
(±)-2-Aminobutyric acid
Vzorec: C₄H₉NO₂
MW: 103,12 g/mol
MDL Number: MFCD00008093
CAS číslo: 2835-81-6
EINECS: 220-616-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR