Ethyl vinyl ketone ≥97% stabilizovaný
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-Penten-3-one
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Stabilized
Purity: 97%
CAS No.: 1629-58-9
Molecular Formula: C5H8O
Formula Weight: 84.12
Storage Temperature: 2°C to 8°C
Flash Point: -6°(21°F)
Refractive Index: 1.4200
Odour: Pungent
Boiling Point: 103-104°
Beilstein Reference: 1735857
UN No.: 3286
MDL No.: MFCD00009316
Vzorec:
C₅H₈O MW: 84,12 g/mol Bod varu: 103…104 °C Hustota: 0,844 Bod vzplanutí: –6 °C (21 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009316 CAS číslo: 1629-58-9 EINECS: 216-624-3 UN: 3286 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...