Tisk…

4-Bromobutyryl chloride ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12094.06 A12094.14 A12094.22
A12094.06EA 1130 CZK
A12094.06 A12094.14 A12094.22
4-Bromobutyryl chloride ≥97%
4-Bromobutyryl chloride

Purity: 97% CAS No.: 927-58-2 Molecular Formula: C4H6BrClO Formula Weight: 185.45 Flash Point: 91°(195°F) Refractive Index: 1.4920 Boiling Point: 102-103°/40mm Beilstein Reference: 1743026 UN No.: 3265 MDL No.: MFCD00000753

Vzorec: C₄H₆BrClO
Bod varu: 102…103 °C (40 mmHg)
Hustota: 1,600
Bod vzplanutí: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000753
CAS číslo: 927-58-2
EINECS: 213-154-0
UN: 3265
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR