4-Bromobutyryl chloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
251933-25G
251933-5G
SIAL251933-25GEA
3690
CZK
SIAL251933-25G
SIAL251933-5G
4-Bromobutyryl chloride, Sigma-Aldrich®
4-Bromobutyryl chloride
Vzorec:
C₄H₆BrClO |
MDL Number:
MFCD00000753 CAS číslo: 927-58-2 EINECS: 213-154-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...