1-Ethyl-4-piperidone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-Ethylpiperidin-4-one
M-Clarity™ quality level = MQ100
279501-25G
279501-100G
SIAL279501-25GEA
2400
CZK
SIAL279501-25G
SIAL279501-100G
1-Ethyl-4-piperidone, Sigma-Aldrich®
1-Ethyl-4-piperidone
Vzorec:
C₇H₁₃NO MW: 127.19 |
MDL Number:
MFCD00013356 CAS číslo: 3612-18-8 EINECS: 222-781-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...