Mesityloxid ≥90% remainder 4-methyl-4-penten-2-one
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Methylpent-3-en-2-on
Purity: 90+%
CAS No.: 141-79-7
Molecular Formula: C6H10O
Formula Weight: 98.15
Flash Point: 30°(86°F)
Physical Form: Clear liquid
Density: 0.856
Refractive Index: 1.4440
Appearance: Colourless
Odour: honigähnlich
Melting Point: -59°
Boiling Point: 129-130°
Beilstein Reference: 1361550
Merck Reference: 14,5908
UN No.: 1229
MDL No.: MFCD00008900
Vzorec:
C₆H₁₀O MW: 98,14 g/mol Bod varu: 129…130 °C Bod tání: –59 °C Hustota: 0,856 Bod vzplanutí: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008900 CAS číslo: 141-79-7 EINECS: 205-502-5 UN: 1229 ADR: 3,III Merck Index: 14,5908 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...