Mesityloxid, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
4-Methylpent-3-en-2-on
M-Clarity™ quality level = MQ300
W336807-1KG
W336807-4KG
W336807-SAMPLE
SIALW336807-1KGEA
0
CZK
SIALW336807-1KG
SIALW336807-4KG
SIALW336807-SAMPLE
Mesityloxid, Sigma-Aldrich®
Mesityloxid
Vzorec:
C₆H₁₀O MW: 98,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008900 CAS číslo: 141-79-7 EINECS: 205-502-5 UN: 1229 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...