2,6-Dimethoxyphenol ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B24253.14
B24253.22
B24253.14EA
1300
CZK
B24253.14
B24253.22
2,6-Dimethoxyphenol ≥99%
2,6-Dimethoxyphenol
Purity: 99%
CAS No.: 91-10-1
Molecular Formula: C8H10O3
Formula Weight: 154.16
Flash Point: 140°(284°F)
Melting Point: 50-57°
Boiling Point: 261°
Beilstein Reference: 1526871
MDL No.: MFCD00064434
Vzorec:
C₈H₁₀O₃ Bod varu: 261 °C Bod tání: 50…57 °C Bod vzplanutí: 140 °C (284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064434 CAS číslo: 91-10-1 EINECS: 202-041-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...