2-Aminocyclohexanol
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR21260-25G
APOSOR21260-25GEA
3580
CZK
APOSOR21260-25G
2-Aminocyclohexanol
2-Aminocyclohexanol
Vzorec:
C₆H₁₃NO MW: 115,18 g/mol |
MDL Number:
MFCD00191368 CAS číslo: 6850-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...