Tisk…

Dihydrogenfosforečnan sodný monohydrát, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

S3522-1KG S3522-250G S3522-500G
SIALS3522-1KGEA 23210 CZK
SIALS3522-1KG SIALS3522-250G SIALS3522-500G
Dihydrogenfosforečnan sodný monohydrát, Sigma-Aldrich®
Dihydrogenfosforečnan sodný monohydrát
Vzorec: NaH₂PO₄·H₂O
MW: 137.99
Bod tání: 100
Hustota: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS číslo: 10049-21-5
EINECS: 231-449-2
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR