Tisk…

β-Glycerophosphate disodium salt hydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Disodium-β-glycerophosphate hydrate, Glycerol 2-phosphate disodium salt hydrate, Sodium 1,3-dihydroxypropan-2-yl phosphate hydrate(2:1:x), BGP

G9422-500G G9422-50G
SIALG9422-500GEA 50080 CZK
SIALG9422-500G SIALG9422-50G
β-Glycerophosphate disodium salt hydrate, Sigma-Aldrich®
β-Glycerophosphate disodium salt hydrate
Vzorec: C₃H₇Na₂O₆P·H₂O
MW: 234,05 g/mol
MDL Number: MFCD00149082
CAS číslo: 154804-51-0
EINECS: 212-464-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR