1-Ethyl-3-methylimidazolium dicyanamide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
EMIM DCA
713384-100G
713384-1KG
SIAL713384-100GEA
13390
CZK
SIAL713384-100G
SIAL713384-1KG
1-Ethyl-3-methylimidazolium dicyanamide, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium dicyanamide
Vzorec:
C₈H₁₁N₅ MW: 177.21 |
MDL Number:
MFCD08276373 CAS číslo: 370865-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...