Bromoacetaldehyde dimethyl acetal ≥97% stabilizovaný
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Bromo-1,1-dimethoxyethane
Stabilization:
Stabilised with potassium carbonate
A11324.0B
A11324.14
A11324.30
A11324.0BEA
23510
CZK
A11324.0B
A11324.14
A11324.30
Bromoacetaldehyde dimethyl acetal ≥97% stabilizovaný
Bromoacetaldehyde dimethyl acetal
Stabilized with potassium carbonate
Purity: 97+%
CAS No.: 7252-83-7
Molecular Formula: C6H13BrO2
Formula Weight: 169.03
Storage Temperature: 2°C to 8°C
Flash Point: 54°(129°F)
Refractive Index: 1.4450
Boiling Point: 150°
Beilstein Reference: 1280132
UN No.: 2929
MDL No.: MFCD00000213
Vzorec:
BrH₂CCH(OCH₃)₂ MW: 169,02 g/mol Bod varu: 150 °C Hustota: 1,435 Bod vzplanutí: 54 °C (129 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000213 CAS číslo: 7252-83-7 EINECS: 230-669-6 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...