Tisk…

L(-)-Xylose, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

851590-25G 851590-5G
SIAL851590-25GEA 7530 CZK
SIAL851590-25G SIAL851590-5G
L(-)-Xylose, Sigma-Aldrich®
L(-)-Xylose
Vzorec: C₅H₁₀O₅
Bod varu: 331 °C (1013 hPa)
Bod tání: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00151096
CAS číslo: 609-06-3
EINECS: 210-174-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR