2-Amino-5-methylpyridine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
6-Amino-3-picoline, 5-Methyl-2-pyridylamine, 5-Methylpyridin-2-amine, 2-Amino-5-picoline
M-Clarity™ quality level = MQ200
A75684-10G
A75684-50G
SIALA75684-10GEA
1290
CZK
SIALA75684-10G
SIALA75684-50G
2-Amino-5-methylpyridine, Sigma-Aldrich®
2-Amino-5-methylpyridine
Vzorec:
C₆H₈N₂ MW: 108.14 Bod varu: 227.1 Bod tání: 74…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006328 CAS číslo: 1603-41-4 EINECS: 216-503-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...