Tisk…

3-Methyladenine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Methyl-3H-purin-6-amine, 3-Methyl-3H-adenine

M9281-100MG M9281-500MG
SIALM9281-100MGEA 6420 CZK
SIALM9281-100MG SIALM9281-500MG
3-Methyladenine, Sigma-Aldrich®
3-Methyladenine
Vzorec: C₆H₇N₅
MDL Number: MFCD00010531
CAS číslo: 5142-23-4
EINECS: 225-908-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR