Guanidiniumthiokyanát, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide
G6639-1KG
G6639-250G
G6639-25G
G6639-500G
G6639-100G
SIALG6639-1KGEA
19970
CZK
SIALG6639-1KG
SIALG6639-250G
SIALG6639-25G
SIALG6639-500G
SIALG6639-100G
Guanidiniumthiokyanát, Sigma-Aldrich®
Guanidiniumthiokyanát
Vzorec:
CH₅N₃·CHNS Bod tání: 118 °C Hustota: 1,29 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013027 CAS číslo: 593-84-0 EINECS: 209-812-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...