Tisk…

Manganese(II) chloride, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Manganese dichloride

M-Clarity™ quality level = MQ200

M1787-100ML M1787-10X1ML
SIALM1787-100MLEA 5900 CZK
SIALM1787-100ML SIALM1787-10X1ML
Manganese(II) chloride, Sigma-Aldrich®
Manganese(II) chloride
Vzorec: MnCl₂
MW: 125.84
Bod varu: 1190
Bod tání: 650
Hustota: 2.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS číslo: 7773-01-5
EINECS: 231-869-6
UN: 3288
ADR: 6.1 III
Merck Index: 12,05768

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR