Manganese(II) chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Manganese dichloride
M-Clarity™ quality level = MQ100
328146-25G
328146-2KG
328146-500G
SIAL328146-25GEA
1090
CZK
SIAL328146-25G
SIAL328146-2KG
SIAL328146-500G
Manganese(II) chloride, Sigma-Aldrich®
Manganese(II) chloride
Vzorec:
MnCl₂ Bod varu: 1190 °C (1013 hPa) Bod tání: 650 °C Hustota: 2,98 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011114 CAS číslo: 7773-01-5 EINECS: 231-869-6 UN: 3288 ADR: 6.1,III Merck Index: 12,05768 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...