Hydrochinonmonomethylether, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ200
M18655-1KG
M18655-100G
M18655-500G
M18655-5G
SIALM18655-1KGEA
4240
CZK
SIALM18655-1KG
SIALM18655-100G
SIALM18655-500G
SIALM18655-5G
Hydrochinonmonomethylether, Sigma-Aldrich®
Hydrochinonmonomethylether
Vzorec:
C₇H₈O₂ Bod varu: 243 °C (1013 hPa) Bod tání: 55…57 °C Hustota: 1,55 g/cm³ (20 °C) Bod vzplanutí: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002332 CAS číslo: 150-76-5 EINECS: 205-769-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...